Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCTTTTGACCATAAGTCACATA[C/G]AATTGTGATTAGGAAATCAATTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602126 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZBTB14 - zinc finger and BTB domain containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143823.2 | 1905 | UTR 3 | NP_001137295.1 | |||
NM_001243702.1 | 1905 | UTR 3 | NP_001230631.1 | |||
NM_001243704.1 | 1905 | UTR 3 | NP_001230633.1 | |||
NM_003409.4 | 1905 | UTR 3 | NP_003400.2 |