Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGGCAGCGGCCACACCATCCA[A/G]GTGAGGGCCTCGGAGAGAGCCGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600805 | ||||||||||||||||||||
Literature Links: |
LAMA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LAMA3 - laminin subunit alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000227.4 | 535 | Intron | NP_000218.3 | |||
NM_001127717.2 | 535 | Silent Mutation | CAA,CAG | Q,Q 98 | NP_001121189.2 | |
NM_001127718.2 | 535 | Intron | NP_001121190.2 | |||
NM_001302996.1 | 535 | Silent Mutation | CAA,CAG | Q,Q 98 | NP_001289925.1 | |
NM_198129.2 | 535 | Silent Mutation | CAA,CAG | Q,Q 98 | NP_937762.2 | |
XM_011525978.2 | 535 | Silent Mutation | CAA,CAG | Q,Q 98 | XP_011524280.1 | |
XM_011525979.2 | 535 | Silent Mutation | CAA,CAG | Q,Q 98 | XP_011524281.1 | |
XM_011525980.2 | 535 | Silent Mutation | CAA,CAG | Q,Q 98 | XP_011524282.1 | |
XM_011525981.2 | 535 | Silent Mutation | CAA,CAG | Q,Q 98 | XP_011524283.1 | |
XM_011525982.2 | 535 | Silent Mutation | CAA,CAG | Q,Q 98 | XP_011524284.1 | |
XM_017025743.1 | 535 | Intron | XP_016881232.1 | |||
XM_017025744.1 | 535 | Intron | XP_016881233.1 |