Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCGAGCGGCCTGGCAGCAGCTC[C/T]GAAACCTTGTCCTCATGCCTGAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603684 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIPG - lipase G, endothelial type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308006.1 | Intron | NP_001294935.1 | ||||
NM_006033.3 | Intron | NP_006024.1 | ||||
XM_005258390.1 | Intron | XP_005258447.1 | ||||
XM_011526265.2 | Intron | XP_011524567.1 | ||||
XM_011526267.1 | Intron | XP_011524569.1 | ||||
XM_017026095.1 | Intron | XP_016881584.1 |