Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTCCAAGAATGAGCAAAGGCAT[A/G]CTGGTAGAAAATCATTGTGCTTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606792 MIM: 615319 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRH4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HRH4 - histamine receptor H4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143828.1 | Intron | NP_001137300.1 | ||||
NM_001160166.1 | Intron | NP_001153638.1 | ||||
NM_021624.3 | Intron | NP_067637.2 | ||||
XM_011526133.1 | Intron | XP_011524435.1 |
IMPACT - impact RWD domain protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |