Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACTCCAAAACTGTACCCAAAACA[C/T]GTGGAGGTATGGGTTCATATTTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR87 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR87 - WD repeat domain 87 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291088.1 | 7717 | Missense Mutation | CAT,CGT | H,R 2488 | NP_001278017.1 | |
NM_031951.4 | 7717 | Missense Mutation | CAT,CGT | H,R 2449 | NP_114157.4 | |
XM_011527360.2 | 7717 | Missense Mutation | CAT,CGT | H,R 2488 | XP_011525662.1 |