Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTCGCAGGGCCCGCCTGGAGCC[A/T]GCTGGCGCGCGTGGCTGGCAGGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602768 MIM: 607381 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLL3 - delta like canonical Notch ligand 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016941.3 | 483 | Missense Mutation | CAG,CTG | Q,L 142 | NP_058637.1 | |
NM_203486.2 | 483 | Missense Mutation | CAG,CTG | Q,L 142 | NP_982353.1 |
TIMM50 - translocase of inner mitochondrial membrane 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |