Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAACGGGAGAAGCCAGAGACAC[A/G]AGAGAGAGGCCCCTGGAAACAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600283 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIK5 - glutamate ionotropic receptor kainate type subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301030.1 | Intron | NP_001287959.1 | ||||
NM_002088.4 | Intron | NP_002079.3 | ||||
XM_005258821.3 | Intron | XP_005258878.1 | ||||
XM_011526862.2 | Intron | XP_011525164.1 | ||||
XM_011526863.2 | Intron | XP_011525165.1 | ||||
XM_011526865.1 | Intron | XP_011525167.1 | ||||
XM_011526867.1 | Intron | XP_011525169.1 | ||||
XM_011526868.1 | Intron | XP_011525170.1 | ||||
XM_011526869.2 | Intron | XP_011525171.1 | ||||
XM_011526870.2 | Intron | XP_011525172.1 | ||||
XM_011526871.2 | Intron | XP_011525173.1 | ||||
XM_017026713.1 | Intron | XP_016882202.1 |
ZNF574 - zinc finger protein 574 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022752.5 | Intron | NP_073589.4 | ||||
XM_005259159.2 | Intron | XP_005259216.1 | ||||
XM_011527228.2 | Intron | XP_011525530.1 | ||||
XM_011527229.2 | Intron | XP_011525531.1 | ||||
XM_011527230.2 | Intron | XP_011525532.1 | ||||
XM_017027149.1 | Intron | XP_016882638.1 |