Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCCCCACCAGGAAAACTGGTGA[A/C]AAGGACCTACATGATGCTGGCTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613176 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMG9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMG9 - SMG9, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005259057.2 | Intron | XP_005259114.1 | ||||
XM_011527113.1 | Intron | XP_011525415.1 | ||||
XM_011527114.1 | Intron | XP_011525416.1 | ||||
XM_011527115.1 | Intron | XP_011525417.1 | ||||
XM_011527116.1 | Intron | XP_011525418.1 | ||||
XM_017026988.1 | Intron | XP_016882477.1 |