Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTAGCTCATCCATTTATCCATTC[A/C]TCATAATCCAGTGTCCAAAGAGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109190 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC1A5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC1A5 - solute carrier family 1 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145144.1 | 1359 | UTR 3 | NP_001138616.1 | |||
NM_001145145.1 | 1359 | UTR 3 | NP_001138617.1 | |||
NM_005628.2 | 1359 | UTR 3 | NP_005619.1 | |||
XM_005259167.4 | 1359 | Intron | XP_005259224.1 |