Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAATAGGGTGGGTGGTCTTTGGC[A/T]GTGGGTAGGGCTGAGGCCTTGACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605890 MIM: 605691 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EHD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EHD2 - EH domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GLTSCR2 - glioma tumor suppressor candidate region gene 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015710.4 | Intron | NP_056525.2 |
GLTSCR2-AS1 - GLTSCR2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD23 - small nucleolar RNA, C/D box 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |