Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTAGCGGCAGATGTTGGTATAGA[T/C]GCCAGGTTTGTCGGACCTCCCACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605644 MIM: 605504 | ||||||||||||||||||||
Literature Links: |
KLK8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLK8 - kallikrein related peptidase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281431.1 | 477 | Missense Mutation | ATC,GTC | I,V 120 | NP_001268360.1 | |
NM_007196.3 | 477 | Missense Mutation | ATC,GTC | I,V 241 | NP_009127.1 | |
NM_144505.2 | 477 | Missense Mutation | ATC,GTC | I,V 286 | NP_653088.1 | |
NM_144506.2 | 477 | Missense Mutation | ATC,GTC | I,V 100 | NP_653089.1 | |
NM_144507.2 | 477 | UTR 3 | NP_653090.1 |
KLK9 - kallikrein related peptidase 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |