Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCTCCTTCATTCCTGTGCTCAA[A/G]CATCACTAACAGAAACACAAGTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606094 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEACAM18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEACAM18 - carcinoembryonic antigen related cell adhesion molecule 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIGLEC12 - sialic acid binding Ig like lectin 12 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033329.2 | Intron | NP_201586.1 | ||||
NM_053003.3 | Intron | NP_443729.1 | ||||
XM_011527441.2 | Intron | XP_011525743.1 | ||||
XM_011527442.2 | Intron | XP_011525744.1 |