Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCCTGGCTGCCCTGACTTCTTT[C/T]GAAGCTCCTTTTAAACCCTTTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606043 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF331 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF331 - zinc finger protein 331 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079906.1 | Intron | NP_001073375.1 | ||||
NM_001079907.1 | Intron | NP_001073376.1 | ||||
NM_001253798.1 | Intron | NP_001240727.1 | ||||
NM_001253799.1 | Intron | NP_001240728.1 | ||||
NM_001253800.1 | Intron | NP_001240729.1 | ||||
NM_001253801.2 | Intron | NP_001240730.1 | ||||
NM_001317113.1 | Intron | NP_001304042.1 | ||||
NM_001317114.1 | Intron | NP_001304043.1 | ||||
NM_001317115.1 | Intron | NP_001304044.1 | ||||
NM_001317116.1 | Intron | NP_001304045.1 | ||||
NM_001317117.1 | Intron | NP_001304046.1 | ||||
NM_001317118.1 | Intron | NP_001304047.1 | ||||
NM_001317119.1 | Intron | NP_001304048.1 | ||||
NM_001317120.1 | Intron | NP_001304049.1 | ||||
NM_001317121.1 | Intron | NP_001304050.1 | ||||
NM_018555.5 | Intron | NP_061025.5 | ||||
XM_011527076.2 | Intron | XP_011525378.1 | ||||
XM_011527078.2 | Intron | XP_011525380.1 | ||||
XM_017026936.1 | Intron | XP_016882425.1 | ||||
XM_017026937.1 | Intron | XP_016882426.1 | ||||
XM_017026938.1 | Intron | XP_016882427.1 | ||||
XM_017026939.1 | Intron | XP_016882428.1 | ||||
XM_017026940.1 | Intron | XP_016882429.1 |