Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTTTGAGTAAGGGCTAAATAAC[C/T]GCTGAAAGCTTTTCCCTATTCTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613905 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf18 - chromosome 19 open reading frame 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF606 - zinc finger protein 606 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025027.3 | 3039 | UTR 3 | NP_079303.2 | |||
XM_005259276.1 | 3039 | UTR 3 | XP_005259333.1 |