Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCCCATCCCTGCAAATCCAGGC[G/A]TCCCCCCGCTGCTGGTCAGACACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142440 MIM: 600235 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HPN - hepsin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002151.2 | 582 | Intron | NP_002142.1 | |||
NM_182983.2 | 582 | UTR 5 | NP_892028.1 | |||
XM_005258838.4 | 582 | Intron | XP_005258895.2 | |||
XM_006723181.3 | 582 | Intron | XP_006723244.2 | |||
XM_017026731.1 | 582 | UTR 5 | XP_016882220.1 | |||
XM_017026732.1 | 582 | Intron | XP_016882221.1 |
SCN1B - sodium voltage-gated channel beta subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |