Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTAGAAATAACACAGGAGAGAC[A/G]CCCATTTCCAACTCTAGGAATCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01534 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01534 - long intergenic non-protein coding RNA 1534 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF567 - zinc finger protein 567 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300979.1 | Intron | NP_001287908.1 | ||||
NM_001322911.1 | Intron | NP_001309840.1 | ||||
NM_001322912.1 | Intron | NP_001309841.1 | ||||
NM_001322913.1 | Intron | NP_001309842.1 | ||||
NM_001322914.1 | Intron | NP_001309843.1 | ||||
NM_001322915.1 | Intron | NP_001309844.1 | ||||
NM_001322916.1 | Intron | NP_001309845.1 | ||||
NM_001322917.1 | Intron | NP_001309846.1 | ||||
NM_001322918.1 | Intron | NP_001309847.1 | ||||
NM_001322919.1 | Intron | NP_001309848.1 | ||||
NM_001322920.1 | Intron | NP_001309849.1 | ||||
NM_152603.4 | Intron | NP_689816.2 | ||||
XM_011526584.2 | Intron | XP_011524886.1 | ||||
XM_011526585.2 | Intron | XP_011524887.1 | ||||
XM_017026416.1 | Intron | XP_016881905.1 | ||||
XM_017026417.1 | Intron | XP_016881906.1 | ||||
XM_017026418.1 | Intron | XP_016881907.1 | ||||
XM_017026419.1 | Intron | XP_016881908.1 | ||||
XM_017026420.1 | Intron | XP_016881909.1 | ||||
XM_017026421.1 | Intron | XP_016881910.1 |