Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGAGGATTTTTCTACATTGATT[A/T]TGGTCCCATAATTTCTCTCCTGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF585A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF585A - zinc finger protein 585A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288800.1 | 571 | Missense Mutation | CAA,CAT | Q,H 104 | NP_001275729.1 | |
NM_152655.3 | 571 | Missense Mutation | CAA,CAT | Q,H 49 | NP_689868.1 | |
NM_199126.2 | 571 | Missense Mutation | CAA,CAT | Q,H 49 | NP_954577.1 |