Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTGTTTGGTTACCCTGAAGGACA[A/G]TTCATATAGGAAAGAAGAATGTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF383 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF383 - zinc finger protein 383 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152604.1 | Intron | NP_689817.1 | ||||
XM_005258585.2 | Intron | XP_005258642.1 | ||||
XM_005258587.2 | Intron | XP_005258644.1 | ||||
XM_005258588.2 | Intron | XP_005258645.1 | ||||
XM_011526586.2 | Intron | XP_011524888.1 | ||||
XM_011526587.2 | Intron | XP_011524889.1 | ||||
XM_011526588.2 | Intron | XP_011524890.1 | ||||
XM_011526589.2 | Intron | XP_011524891.1 | ||||
XM_011526590.2 | Intron | XP_011524892.1 | ||||
XM_017026422.1 | Intron | XP_016881911.1 | ||||
XM_017026423.1 | Intron | XP_016881912.1 | ||||
XM_017026424.1 | Intron | XP_016881913.1 |