Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGAAGAAGAAGTTGCGAAGGAT[A/C]CCCAACGGGGAGGCCAACGTGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602216 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STK11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STK11 - serine/threonine kinase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000455.4 | 889 | Silent Mutation | ATA,ATC | I,I 88 | NP_000446.1 | |
XM_005259617.2 | 889 | Silent Mutation | ATA,ATC | I,I 88 | XP_005259674.1 | |
XM_005259618.3 | 889 | Silent Mutation | ATA,ATC | I,I 88 | XP_005259675.1 | |
XM_011528209.1 | 889 | UTR 5 | XP_011526511.1 |