Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGGTGAGCTGTCCCCACCGTGA[C/T]TCAGAAGGTATTTTCTGCTGCTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607375 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOT1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOT1L - DOT1 like histone lysine methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032482.2 | Intron | NP_115871.1 | ||||
XM_005259659.3 | Intron | XP_005259716.1 | ||||
XM_005259660.3 | Intron | XP_005259717.1 | ||||
XM_011528359.2 | Intron | XP_011526661.1 | ||||
XM_011528360.1 | Intron | XP_011526662.1 | ||||
XM_011528361.2 | Intron | XP_011526663.1 | ||||
XM_017027366.1 | Intron | XP_016882855.1 |