Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTAGGGGGAAGGACCAGAGGGC[A/G]TGCCTCTAGCAGAGCTAGGAGCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612680 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CELF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CELF5 - CUGBP, Elav-like family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172673.1 | Intron | NP_001166144.1 | ||||
NM_021938.3 | Intron | NP_068757.2 | ||||
XM_006722832.1 | Intron | XP_006722895.1 | ||||
XM_006722833.1 | Intron | XP_006722896.1 | ||||
XM_006722836.2 | Intron | XP_006722899.1 | ||||
XM_006722837.3 | Intron | XP_006722900.1 | ||||
XM_006722838.3 | Intron | XP_006722901.1 | ||||
XM_011528173.1 | Intron | XP_011526475.1 | ||||
XM_011528174.2 | Intron | XP_011526476.1 | ||||
XM_017027111.1 | Intron | XP_016882600.1 |