Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCCTGGTGCAGGATCACGCCCA[A/G]GAGACGCACCAGGTTCTCGTGTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600038 MIM: 610362 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MATK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MATK - megakaryocyte-associated tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002378.3 | 1274 | Silent Mutation | CTG,TTG | L,L 293 | NP_002369.2 | |
NM_139354.2 | 1274 | Silent Mutation | CTG,TTG | L,L 251 | NP_647611.1 | |
NM_139355.2 | 1274 | Silent Mutation | CTG,TTG | L,L 292 | NP_647612.1 | |
XM_011528018.1 | 1274 | Silent Mutation | CTG,TTG | L,L 292 | XP_011526320.1 |
RAX2 - retina and anterior neural fold homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |