Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGGCTTTTCTCCAGTGTGAATT[C/G]GCATGTGTAAAATAAGTCCTGTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF846 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF846 - zinc finger protein 846 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077624.1 | 1960 | Missense Mutation | CCA,CGA | P,R 330 | NP_001071092.1 | |
XM_005259772.3 | 1960 | Missense Mutation | CCA,CGA | P,R 330 | XP_005259829.1 | |
XM_005259773.4 | 1960 | Missense Mutation | CCA,CGA | P,R 329 | XP_005259830.1 | |
XM_005259775.3 | 1960 | Missense Mutation | CCA,CGA | P,R 329 | XP_005259832.1 | |
XM_006722658.3 | 1960 | Missense Mutation | CCA,CGA | P,R 330 | XP_006722721.1 | |
XM_011527717.2 | 1960 | Missense Mutation | CCA,CGA | P,R 330 | XP_011526019.1 | |
XM_011527718.2 | 1960 | Missense Mutation | CCA,CGA | P,R 329 | XP_011526020.1 | |
XM_011527719.2 | 1960 | Missense Mutation | CCA,CGA | P,R 294 | XP_011526021.1 | |
XM_011527720.2 | 1960 | Missense Mutation | CCA,CGA | P,R 294 | XP_011526022.1 | |
XM_011527721.2 | 1960 | Missense Mutation | CCA,CGA | P,R 273 | XP_011526023.1 | |
XM_017026404.1 | 1960 | Missense Mutation | CCA,CGA | P,R 316 | XP_016881893.1 | |
XM_017026405.1 | 1960 | Missense Mutation | CCA,CGA | P,R 201 | XP_016881894.1 | |
XM_017026406.1 | 1960 | Missense Mutation | CCA,CGA | P,R 201 | XP_016881895.1 | |
XM_017026407.1 | 1960 | Missense Mutation | CCA,CGA | P,R 330 | XP_016881896.1 | |
XM_017026408.1 | 1960 | Missense Mutation | CCA,CGA | P,R 329 | XP_016881897.1 |