Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACCAAGGCTGACTCACCTCCCA[G/T]CCGGAACTCTGTGGGGAAGAGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614194 MIM: 614610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOCK6 - dedicator of cytokinesis 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020812.3 | 5323 | Silent Mutation | NP_065863.2 | |||
XM_005260000.2 | 5323 | Silent Mutation | XP_005260057.1 | |||
XM_005260001.2 | 5323 | Silent Mutation | XP_005260058.1 | |||
XM_006722804.3 | 5323 | Silent Mutation | XP_006722867.1 | |||
XM_011528150.1 | 5323 | Silent Mutation | XP_011526452.1 | |||
XM_011528151.1 | 5323 | Silent Mutation | XP_011526453.1 | |||
XM_011528152.1 | 5323 | Silent Mutation | XP_011526454.1 |
KANK2 - KN motif and ankyrin repeat domains 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105372273 - uncharacterized LOC105372273 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |