Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGCTGAAGGTGAAGCAGGGGGC[A/G]GAGAACATGATCCAGACCTACAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601032 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PKN1 - protein kinase N1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002741.3 | 470 | Silent Mutation | GCA,GCG | A,A 145 | NP_002732.3 | |
NM_213560.1 | 470 | Silent Mutation | GCA,GCG | A,A 151 | NP_998725.1 | |
XM_011528128.1 | 470 | Silent Mutation | GCA,GCG | A,A 157 | XP_011526430.1 |