Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGAATTCCGAAGCAGAAGCCAG[A/C]GCCCCACAGGAGCTAACGGCCAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600317 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TPM4 - tropomyosin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145160.1 | Intron | NP_001138632.1 | ||||
NM_003290.2 | Intron | NP_003281.1 | ||||
XM_005260042.3 | Intron | XP_005260099.1 | ||||
XM_006722865.2 | Intron | XP_006722928.1 | ||||
XM_011528231.2 | Intron | XP_011526533.1 | ||||
XM_011528232.1 | Intron | XP_011526534.1 | ||||
XM_017027188.1 | Intron | XP_016882677.1 | ||||
XM_017027189.1 | Intron | XP_016882678.1 |