Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCTCGCCCCACAGTACAGCATA[A/C]AACCAGTAGCACCCACAATAACTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605043 | ||||||||||||||||||||
Literature Links: |
MED26 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MED26 - mediator complex subunit 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004831.3 | 2923 | UTR 3 | NP_004822.2 |
SLC35E1 - solute carrier family 35 member E1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |