Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATATTATTCAGGGACAAAATA[A/G]AGATTTCCAGTCCCCATGTATGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602967 | ||||||||||||||||||||
Literature Links: |
LOC101927770 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927770 - uncharacterized LOC101927770 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF217 - zinc finger protein 217 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006526.2 | 3772 | UTR 3 | NP_006517.1 | |||
XM_005260545.3 | 3772 | UTR 3 | XP_005260602.1 | |||
XM_006723875.2 | 3772 | UTR 3 | XP_006723938.1 | |||
XM_011529036.1 | 3772 | UTR 3 | XP_011527338.1 | |||
XM_017028059.1 | 3772 | UTR 3 | XP_016883548.1 |