Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGCCAATATCAAGTGAATGAAT[C/T]CATAGGTTTGAGGTGTTACCCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607022 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTCFL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTCFL - CCCTC-binding factor like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001269040.1 | 2803 | UTR 3 | NP_001255969.1 | |||
NM_001269041.1 | 2803 | UTR 3 | NP_001255970.1 | |||
NM_001269042.1 | 2803 | UTR 3 | NP_001255971.1 | |||
NM_001269043.1 | 2803 | Intron | NP_001255972.1 | |||
NM_001269044.1 | 2803 | Intron | NP_001255973.1 | |||
NM_001269045.1 | 2803 | Intron | NP_001255974.1 | |||
NM_001269046.1 | 2803 | UTR 3 | NP_001255975.1 | |||
NM_001269047.1 | 2803 | Intron | NP_001255976.1 | |||
NM_001269048.1 | 2803 | Intron | NP_001255977.1 | |||
NM_001269049.1 | 2803 | Intron | NP_001255978.1 | |||
NM_001269050.1 | 2803 | Intron | NP_001255979.1 | |||
NM_001269051.1 | 2803 | Intron | NP_001255980.1 | |||
NM_001269052.1 | 2803 | Intron | NP_001255981.1 | |||
NM_001269054.1 | 2803 | Intron | NP_001255983.1 | |||
NM_001269055.1 | 2803 | Intron | NP_001255984.1 | |||
NM_080618.3 | 2803 | UTR 3 | NP_542185.2 |