Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATGCTGCCGAGCTCTGGAAAAA[A/C]CCCACAGGTGAGTCCTCAGTCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611388 MIM: 191039 MIM: 605574 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNTTIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNTTIP1 - deoxynucleotidyltransferase, terminal, interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNNC2 - troponin C2, fast skeletal type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE2C - ubiquitin conjugating enzyme E2 C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281741.1 | 631 | Intron | NP_001268670.1 | |||
NM_001281742.1 | 631 | Intron | NP_001268671.1 | |||
NM_007019.3 | 631 | Intron | NP_008950.1 | |||
NM_181799.2 | 631 | Intron | NP_861515.1 | |||
NM_181800.2 | 631 | Intron | NP_861516.1 | |||
NM_181801.3 | 631 | Missense Mutation | AAA,AAC | K,N 119 | NP_861517.1 |