Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGACGCTGATTAGCAAGTGCTCC[A/G]ATGGGCAGCCTGCCTTTCCTGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602461 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIRPA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIRPA - signal regulatory protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040022.1 | Intron | NP_001035111.1 | ||||
NM_001040023.1 | Intron | NP_001035112.1 | ||||
NM_080792.2 | Intron | NP_542970.1 | ||||
XM_005260669.3 | Intron | XP_005260726.1 | ||||
XM_005260670.3 | Intron | XP_005260727.1 | ||||
XM_006723545.3 | Intron | XP_006723608.1 | ||||
XM_011529173.2 | Intron | XP_011527475.1 |