Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAAGGACAACAGTTTTACATGG[A/G]AATGAATAGAAAGAAAATACACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610833 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAA20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAA20 - N(alpha)-acetyltransferase 20, NatB catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016100.4 | Intron | NP_057184.1 | ||||
NM_181527.3 | Intron | NP_852668.1 | ||||
NM_181528.3 | Intron | NP_852669.1 |