Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGTTGGATGACAACTTAGATAC[C/T]GAGCGTCCCGTCCAGAAACGAGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607898 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIB3 - tribbles pseudokinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301188.1 | 130 | Silent Mutation | ACC,ACT | T,T 27 | NP_001288117.1 | |
NM_001301190.1 | 130 | Silent Mutation | ACC,ACT | T,T 27 | NP_001288119.1 | |
NM_001301193.1 | 130 | Silent Mutation | ACC,ACT | T,T 27 | NP_001288122.1 | |
NM_001301196.1 | 130 | Silent Mutation | ACC,ACT | T,T 27 | NP_001288125.1 | |
NM_001301201.1 | 130 | Silent Mutation | ACC,ACT | T,T 54 | NP_001288130.1 | |
NM_021158.4 | 130 | Silent Mutation | ACC,ACT | T,T 27 | NP_066981.2 | |
XM_017027989.1 | 130 | Silent Mutation | ACC,ACT | T,T 54 | XP_016883478.1 |