Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCGGGAGGGCGGTGAGCCGGAGA[A/G]AGGCCCCGTCCCCACCCTCGCCACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607114 MIM: 603130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM33 - ADAM metallopeptidase domain 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ATRN - attractin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GFRA4 - GDNF family receptor alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022139.3 | 441 | Intron | NP_071422.1 | |||
NM_145762.2 | 441 | Silent Mutation | CTC,CTT | L,L 147 | NP_665705.1 | |
XM_005260793.2 | 441 | Intron | XP_005260850.1 |