Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTAGTTTCTCCAGGGCTTGGGTT[A/G]ACATTGTAAACTAATGTGGCCTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602621 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXADR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXADR - coxsackie virus and adenovirus receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207063.1 | Intron | NP_001193992.1 | ||||
NM_001207064.1 | Intron | NP_001193993.1 | ||||
NM_001207065.1 | Intron | NP_001193994.1 | ||||
NM_001207066.1 | Intron | NP_001193995.1 | ||||
NM_001338.4 | Intron | NP_001329.1 | ||||
XM_011529476.2 | Intron | XP_011527778.1 | ||||
XM_011529477.2 | Intron | XP_011527779.1 | ||||
XM_011529478.2 | Intron | XP_011527780.1 | ||||
XM_011529479.1 | Intron | XP_011527781.1 |