Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTCCTGAGGGCAGCAGGCCTGG[A/T]TGCAGCTGGCGTGGGGGTCTCAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603749 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRPM2 - transient receptor potential cation channel subfamily M member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320350.1 | 481 | UTR 5 | NP_001307279.1 | |||
NM_001320351.1 | 481 | UTR 5 | NP_001307280.1 | |||
NM_001320352.1 | 481 | Intron | NP_001307281.1 | |||
NM_003307.3 | 481 | UTR 5 | NP_003298.1 | |||
XM_005261171.3 | 481 | UTR 5 | XP_005261228.1 | |||
XM_011529736.2 | 481 | UTR 5 | XP_011528038.1 | |||
XM_017028456.1 | 481 | UTR 5 | XP_016883945.1 | |||
XM_017028457.1 | 481 | UTR 5 | XP_016883946.1 |