Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTTGATTATTTCTAGATATTTC[A/G]TCTTTGGAATCGTCACAGAAGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604702 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGXB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMGXB4 - HMG-box containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003681.2 | 441 | Silent Mutation | TCA,TCG | S,S 89 | NP_001003681.1 | |
XM_006724100.3 | 441 | Silent Mutation | TCA,TCG | S,S 132 | XP_006724163.1 | |
XM_006724101.3 | 441 | Silent Mutation | TCA,TCG | S,S 132 | XP_006724164.1 | |
XM_006724102.1 | 441 | UTR 5 | XP_006724165.1 |