Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTCTAATTTTGTCCAATATTAT[A/G]TATGGAAATTCTCAATGTCGCGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138981 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSF2RB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSF2RB - colony stimulating factor 2 receptor beta common subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000395.2 | Intron | NP_000386.1 | ||||
XM_005261340.3 | Intron | XP_005261397.1 | ||||
XM_011529903.2 | Intron | XP_011528205.1 | ||||
XM_011529904.2 | Intron | XP_011528206.1 | ||||
XM_011529905.2 | Intron | XP_011528207.1 |