Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGCCTATAATATACAGGGTACT[C/T]CATAAATATTGCAGAATGAATACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602721 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DMC1 - DNA meiotic recombinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278208.1 | Intron | NP_001265137.1 | ||||
NM_007068.3 | Intron | NP_008999.2 | ||||
XM_006724112.2 | Intron | XP_006724175.1 | ||||
XM_011529834.1 | Intron | XP_011528136.1 | ||||
XM_011529835.2 | Intron | XP_011528137.1 | ||||
XM_011529837.1 | Intron | XP_011528139.1 | ||||
XM_011529838.1 | Intron | XP_011528140.1 | ||||
XM_017028542.1 | Intron | XP_016884031.1 |