Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGCTGCTGCTGGGTCTGGCCAT[A/G]GTCGCCCATGGCCTGCTGCGCCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147220 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGLC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGLC1 - immunoglobulin lambda constant 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IGLL5 - immunoglobulin lambda like polypeptide 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178126.1 | 373 | Missense Mutation | ATA,ATG | I,M 33 | NP_001171597.1 | |
NM_001256296.1 | 373 | UTR 5 | NP_001243225.1 |
MIR5571 - microRNA 5571 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |