Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGCAGGAGGCTGGAGCTGAGAC[A/G]TGGGTGTTCCTGCAGCTGGGGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608121 MIM: 612058 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARVB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARVB - parvin beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003828.2 | 222 | Missense Mutation | CAT,CGT | H,R 31 | NP_001003828.1 | |
NM_001243385.1 | 222 | Intron | NP_001230314.1 | |||
NM_001243386.1 | 222 | Intron | NP_001230315.1 | |||
NM_013327.4 | 222 | Intron | NP_037459.2 | |||
XM_005261596.1 | 222 | Intron | XP_005261653.1 | |||
XM_017028792.1 | 222 | Intron | XP_016884281.1 |
SAMM50 - SAMM50 sorting and assembly machinery component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |