Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTACGTAAAGGAGTATTGGAAT[A/G]ATGTGTTCAGGTGAGTGCGGGTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607477 MIM: 610230 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTSE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTSE1 - G2 and S-phase expressed 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMU - tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282782.1 | 245 | Missense Mutation | ATA,ATG | I,M 1 | NP_001269711.1 | |
NM_001282783.1 | 245 | UTR 5 | NP_001269712.1 | |||
NM_001282784.1 | 245 | UTR 5 | NP_001269713.1 | |||
NM_001282785.1 | 245 | Missense Mutation | AAT,GAT | N,D 80 | NP_001269714.1 | |
NM_018006.4 | 245 | Missense Mutation | AAT,GAT | N,D 80 | NP_060476.2 | |
XM_005261678.1 | 245 | UTR 5 | XP_005261735.1 | |||
XM_005261681.1 | 245 | UTR 5 | XP_005261738.1 | |||
XM_011530271.2 | 245 | Missense Mutation | AAT,GAT | N,D 45 | XP_011528573.1 | |
XM_011530272.1 | 245 | Missense Mutation | AAT,GAT | N,D 80 | XP_011528574.1 | |
XM_011530273.1 | 245 | Missense Mutation | AAT,GAT | N,D 80 | XP_011528575.1 | |
XM_011530274.2 | 245 | Missense Mutation | ATA,ATG | I,M 1 | XP_011528576.1 | |
XM_011530275.1 | 245 | UTR 5 | XP_011528577.1 |