Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCAGCCAGGAGCTGTCCTCGCC[C/T]GGCTCCGACTCGGAGGACGCGGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607755 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHKB-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHKB-AS1 - CHKB antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK8IP2 - mitogen-activated protein kinase 8 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012324.4 | 782 | Silent Mutation | CCC,CCT | P,P 252 | NP_036456.1 | |
XM_011530679.2 | 782 | Silent Mutation | CCC,CCT | P,P 253 | XP_011528981.1 | |
XM_011530680.2 | 782 | Silent Mutation | CCC,CCT | P,P 215 | XP_011528982.1 | |
XM_011530681.2 | 782 | Silent Mutation | CCC,CCT | P,P 208 | XP_011528983.1 |