Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAACAGCCGCCTGCGTTTCACTAA[C/T]GGGGGTAAGCCTCCCTGTATGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300916 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNMA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNMA5 - paraneoplastic Ma antigen family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001103150.1 | 1503 | Silent Mutation | CCA,CCG | P,P 388 | NP_001096620.1 | |
NM_001103151.1 | 1503 | Silent Mutation | CCA,CCG | P,P 388 | NP_001096621.1 | |
NM_001184924.1 | 1503 | Silent Mutation | CCA,CCG | P,P 388 | NP_001171853.1 | |
NM_052926.2 | 1503 | Silent Mutation | CCA,CCG | P,P 388 | NP_443158.1 | |
XM_017029252.1 | 1503 | Silent Mutation | CCA,CCG | P,P 388 | XP_016884741.1 | |
XM_017029253.1 | 1503 | Silent Mutation | CCA,CCG | P,P 388 | XP_016884742.1 |