Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAATATATAATCAACTGTTGGTG[A/T]GATAACAGGATAAAATTCCACCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300631 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GS1-594A7.3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GS1-594A7.3 - uncharacterized LOC104798195 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM27 - transmembrane protein 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020665.5 | 1443 | UTR 3 | NP_065716.1 | |||
XM_017029680.1 | 1443 | UTR 3 | XP_016885169.1 | |||
XM_017029681.1 | 1443 | UTR 3 | XP_016885170.1 |