Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTGGAGGGGTGACCAGTTCATT[G/T]TAGGGTCCCCATAAGCAGCTAATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300948 MIM: 300906 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCYT1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCYT1B - phosphate cytidylyltransferase 1, choline, beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163264.1 | 4595 | UTR 3 | NP_001156736.1 | |||
NM_001163265.1 | 4595 | UTR 3 | NP_001156737.1 | |||
NM_004845.4 | 4595 | UTR 3 | NP_004836.2 | |||
XM_017029977.1 | 4595 | UTR 3 | XP_016885466.1 |
PDK3 - pyruvate dehydrogenase kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |