Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCTTGTCTCAGGAGACACATTCA[A/G]ATTCTGCAAGAATTTTAATGTCTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300002 | ||||||||||||||||||||
Literature Links: |
ARSD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARSD - arylsulfatase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001669.3 | 1271 | Intron | NP_001660.2 | |||
NM_009589.3 | 1271 | UTR 3 | NP_033667.2 | |||
XM_005274514.2 | 1271 | Intron | XP_005274571.1 | |||
XM_005274515.2 | 1271 | Intron | XP_005274572.1 |