Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGAATACACATTCCAATTAAAAG[A/G]CAAAGACTGACAGAATGTATACAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603442 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDKL2 - cyclin dependent kinase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005263335.2 | Intron | XP_005263392.1 | ||||
XM_006714406.2 | Intron | XP_006714469.1 | ||||
XM_017008809.1 | Intron | XP_016864298.1 | ||||
XM_017008810.1 | Intron | XP_016864299.1 | ||||
XM_017008811.1 | Intron | XP_016864300.1 |