Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGAAAGATATTGATATACGATG[A/G]AACAAAAATCATTTTCTTATTGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612127 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD17B13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSD17B13 - hydroxysteroid 17-beta dehydrogenase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136230.2 | 843 | Missense Mutation | CCA,TCA | P,S 224 | NP_001129702.1 | |
NM_178135.4 | 843 | Missense Mutation | CCA,TCA | P,S 260 | NP_835236.2 |
MIR5705 - microRNA 5705 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |