Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGATGGTCAAAGTTTTGTAACGT[C/T]GGTGTATTGGAACCACACCACAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606022 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUDT9 - nudix hydrolase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001248011.1 | 1130 | Intron | NP_001234940.1 | |||
NM_024047.4 | 1130 | Intron | NP_076952.1 | |||
NM_198038.2 | 1130 | Intron | NP_932155.1 | |||
XM_011532031.2 | 1130 | Intron | XP_011530333.1 | |||
XM_011532032.2 | 1130 | UTR 5 | XP_011530334.1 |